Stampa…
Swift™ stain_membrane
Fornitore: G Biosciences
Pericolo
786-677
786-677EA 489 EUR
786-677
Swift™ stain_membrane
Swift™
Swift™ Membrane Stain is a unique, proprietary (patents pending), rapid, reversible, ready-to-use membrane stain for proteins on nitrocellulose or PVDF membranes. Swift™ Membrane Stain stains proteins faster and with 500 times more sensitivity than the routinely used Ponceau-S stain and other commercially available protein stains.
  • Reversible, ready-to-use membrane stain for proteins
  • Compatible with nitrocellulose or PVDF
  • Lower detection limit is ~0.5ng protein (BSA)/band on nitrocellulose membrane
  • Outperforms routinely used Ponceau-S
  • Outperforms Coomassie based stains
Can be completely removed from the membrane in less than one minute without affecting the biological or immunological properties of the immobilised proteins.
Informazioni per l'ordine: The kit components are sufficient for 20 blots of 8×8cm size.