Stampa…
pET-14b DNA, Novagen®
Fornitore: Merck Millipore (Novagen)
69660-3
69660-3EA 396 EUR
69660-3
pET-14b DNA, Novagen®
Vettori, plasmidi e librerie
The pET-14b vector carries an N-terminal His·Tag® sequence followed by a thrombin cleavage site and three cloning sites.
  • Packed in a plastic ampoule
  • Store at ≤–70 °C
Informazioni per l'ordine: The pET vectors are supplied as purified plasmid DNA (10 µg). It includes an induction control strain (supplied as a glycerol stock).