Stampa…
EZ-Fit® Filtration Unit with Absorbent Pad
Fornitore: Merck
EFAAW100B
MILXEFAAW100BEA 0 EUR
MILXEFAAW100B
EZ-Fit® Filtration Unit with Absorbent Pad
Filtri Filtri per bottiglie
The EZ-FIT® filtration unit is a single-use filtration device made of white, gridded, and mixed cellulose esters (MCE) membrane with a blue base integrating a cellulose support pad. It provides a safe and fast way of performing membrane filtration by avoiding the exposure to the risks of microbiological analysis.
The filtration units are e-beam irrated, a vented lid covers the sample throughout filtration, and a 360° rim prevents forceps from accidentally touching the membrane filtration area during membrane transfer, to protect samples from contamination while ensuring reliable