Stampa…
Kanamycin ELISA Kit - Extracellular
Fornitore: Abcam
AB318950-10X96 AB318950-96TST
ABCAAB318950-10X96EA 8400 EUR
ABCAAB318950-10X96 ABCAAB318950-96TST
Kanamycin ELISA Kit - Extracellular
Saggi ELISA
Kanamycin ELISA Kit - Extracellular is a single-wash 90-min Simplestep used to quantify Kanamycin with a sensitivity of 38,3 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Competitive ELISA - 450 nm readout : works on any standard plate reader - Different formats for different needs: 10×96 plates for bulk orders.