Stampa…
HexArmor® Rig Lizard® Thin Lizzie™ 2090X Safety Gloves, 60648 Series
Fornitore: UVEX
6064809 6064810 6064806 6064807 6064808 6064811
UVEX6064809EA 0 EUR
UVEX6064809 UVEX6064810 UVEX6064806 UVEX6064807 UVEX6064808 UVEX6064811
HexArmor® Rig Lizard® Thin Lizzie™ 2090X Safety Gloves, 60648 Series
Guanti
The Rig Lizard® 2090X by HexArmor® features strong cut and puncture resistance, as well as their hi-vis back-of-hand IR-X® Impact Exoskeleton™ with high-flex design and a reinforced thumb saddle for longer-lasting protection.
  • IR-X® Impact Exoskeleton™ for impact resistance
  • Flexible sandy nitrile palm coating over an HPPE and glass blend shell
  • Hi-vis on back-of-hand enhances visibility and compliance
  • Seamless construction delivers high comfort and dexterity
Back-of-hand IR-X® Impact Exoskeleton™ keeps hands safe from smash and impact hazards. The 13-gauge hi-vis HPPE, and fibreglass shell enhances visibility and worker compliance. Flexible sandy nitrile palm coating provides superior grip and abrasion resistance in wet conditions. Reinforced thumb crotch patch ensures increased durability, and a knit wrist helps prevent dirt and debris from entering the glove. Seamless construction allows enhanced comfort while maintaining dexterity and tactility.