Stampa…

Anti-Cardiac Troponin I Rabbit Monoclonal Antibody [clone: EPR29249-56]

Fornitore: Abcam
AB321922-100UG AB321922-1MG
ABCAAB321922-100UGEA 919 EUR
ABCAAB321922-100UG ABCAAB321922-1MG
Anti-Cardiac Troponin I Rabbit Monoclonal Antibody [clone: EPR29249-56]
Anticorpi
Rabbit monoclonal [EPR29249-56] to Cardiac Troponin I - BSA and Azide free (Detector).

Attenzione: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Cardiac Troponin I
Clonality: Monoclonal
Clone: EPR29249-56
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR