Stampa…

Anti-Zinc finger antiviral protein Rabbit Monoclonal Antibody [clone: EPR28538-63]

Fornitore: Abcam
AB314900-1MG AB314900-100UG
ABCAAB314900-1MGEA 9160 EUR
ABCAAB314900-1MG ABCAAB314900-100UG
Anti-Zinc finger antiviral protein Rabbit Monoclonal Antibody [clone: EPR28538-63]
Anticorpi
Rabbit monoclonal [EPR28538-63] to Zinc finger antiviral protein - BSA and Azide free.

Attenzione: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Zinc finger antiviral protein
Clonality: Monoclonal
Clone: EPR28538-63
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR