Stampa…
Anti-NUT Rabbit Monoclonal Antibody [clone: EPR28221-46]
Fornitore: Abcam
AB315460-20UL AB315460-100UL AB315460-1ML
ABCAAB315460-20ULEA 244 EUR
ABCAAB315460-20UL ABCAAB315460-100UL ABCAAB315460-1ML
Anti-NUT Rabbit Monoclonal Antibody [clone: EPR28221-46]
Anticorpi
Rabbit monoclonal [EPR28221-46] to NUT.
Attenzione: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: NUT
Clonality: Monoclonal
Clone: EPR28221-46
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human