Stampa…
Anti-ST2 Rabbit Monoclonal Antibody [clone: EPR28307-60]
Fornitore: Abcam
AB319146-20UL AB319146-100UL AB319146-1ML
ABCAAB319146-20ULEA 244 EUR
ABCAAB319146-20UL ABCAAB319146-100UL ABCAAB319146-1ML
Anti-ST2 Rabbit Monoclonal Antibody [clone: EPR28307-60]
Anticorpi
Rabbit monoclonal [EPR28307-60] to ST2.
Attenzione: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: ST2
Clonality: Monoclonal
Clone: EPR28307-60
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse