Stampa…

Anti-Tyrosine Hydroxylase Mouse Monoclonal Antibody [clone: EP1532Y]

Fornitore: Abcam
AB317796-100UG AB317796-1MG
ABCAAB317796-100UGEA 1020 EUR
ABCAAB317796-100UG ABCAAB317796-1MG
Anti-Tyrosine Hydroxylase Mouse Monoclonal Antibody [clone: EP1532Y]
Anticorpi
Mouse monoclonal [EP1532Y] to Tyrosine Hydroxylase - Chimeric – BSA and Azide free.

Attenzione: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Tyrosine Hydroxylase
Clonality: Monoclonal
Clone: EP1532Y
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR