Stampa…

Anti-ZCWCC1 Rabbit Monoclonal Antibody [clone: EPR28655-56]

Fornitore: Abcam
AB315903-20UL AB315903-100UL AB315903-1ML
ABCAAB315903-20ULEA 244 EUR
ABCAAB315903-20UL ABCAAB315903-100UL ABCAAB315903-1ML
Anti-ZCWCC1 Rabbit Monoclonal Antibody [clone: EPR28655-56]
Anticorpi
Rabbit monoclonal [EPR28655-56] to ZCWCC1.

Attenzione: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: ZCWCC1
Clonality: Monoclonal
Clone: EPR28655-56
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR