Stampa…
Anti-SGLT2 Rabbit Monoclonal Antibody [clone: EPR29167-46]
Fornitore: Abcam
AB320086-20UL AB320086-100UL AB320086-1ML
ABCAAB320086-20ULEA 244 EUR
ABCAAB320086-20UL ABCAAB320086-100UL ABCAAB320086-1ML
Anti-SGLT2 Rabbit Monoclonal Antibody [clone: EPR29167-46]
Anticorpi
Rabbit monoclonal [EPR29167-46] to SGLT2.
Attenzione: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: SGLT2
Clonality: Monoclonal
Clone: EPR29167-46
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human