Stampa…

Anti-Tyrosine Hydroxylase Chicken Monoclonal Antibody [clone: EP1532Y]

Fornitore: Abcam
AB318304-20UL AB318304-100UL AB318304-1ML
ABCAAB318304-20ULEA 263 EUR
ABCAAB318304-20UL ABCAAB318304-100UL ABCAAB318304-1ML
Anti-Tyrosine Hydroxylase Chicken Monoclonal Antibody [clone: EP1532Y]
Anticorpi
Chicken monoclonal [EP1532Y] to Tyrosine Hydroxylase - Chimeric.

Attenzione: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Tyrosine Hydroxylase
Clonality: Monoclonal
Clone: EP1532Y
Conjugation:
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Mouse;Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR