Stampa…

Anti-LANCL2 Rabbit Monoclonal Antibody [clone: EPR27973-111]

Fornitore: Abcam
AB315946-1MG AB315946-100UG
ABCAAB315946-1MGEA 9200 EUR
ABCAAB315946-1MG ABCAAB315946-100UG
Anti-LANCL2 Rabbit Monoclonal Antibody [clone: EPR27973-111]
Anticorpi
Rabbit monoclonal [EPR27973-111] to LANCL2 - BSA and Azide free (Detector).

Attenzione: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: LANCL2
Clonality: Monoclonal
Clone: EPR27973-111
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR