Stampa…

Anti-Tyrosine Hydroxylase Goat Monoclonal Antibody [clone: EP1532Y]

Fornitore: Abcam
AB317785-20UL AB317785-100UL AB317785-1ML
ABCAAB317785-20ULEA 263 EUR
ABCAAB317785-20UL ABCAAB317785-100UL ABCAAB317785-1ML
Anti-Tyrosine Hydroxylase Goat Monoclonal Antibody [clone: EP1532Y]
Anticorpi
Goat monoclonal [EP1532Y] to Tyrosine Hydroxylase - Chimeric.

Attenzione: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Tyrosine Hydroxylase
Clonality: Monoclonal
Clone: EP1532Y
Conjugation:
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR