Stampa…

Anti-EBV Nuclear Antigen/EBNA1 Rabbit Monoclonal Antibody [clone: EPR28706-78]

Fornitore: Abcam
AB316860-20UL AB316860-100UL AB316860-1ML
ABCAAB316860-20ULEA 244 EUR
ABCAAB316860-20UL ABCAAB316860-100UL ABCAAB316860-1ML
Anti-EBV Nuclear Antigen/EBNA1 Rabbit Monoclonal Antibody [clone: EPR28706-78]
Anticorpi
Rabbit monoclonal [EPR28706-78] to EBV Nuclear Antigen/EBNA1.

Type: Primary
Antigen: EBV Nuclear Antigen/EBNA1
Clonality: Monoclonal
Clone: EPR28706-78
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR