Stampa…

Anti-MEIS2 Rabbit Monoclonal Antibody [clone: EPR12100]

Fornitore: Abcam
AB174270-100 AB174270-1ML
ABCAAB174270-100EA 713 EUR
ABCAAB174270-100 ABCAAB174270-1ML
Anti-MEIS2 Rabbit Monoclonal Antibody [clone: EPR12100]
Anticorpi
Rabbit monoclonal [EPR12100] to MEIS2.

Attenzione: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: MEIS2
Clonality: Monoclonal
Clone: EPR12100
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR