Stampa…
Anti-Cdk9 Rabbit Monoclonal Antibody [clone: EPR22956-60]
Fornitore: Abcam
AB261724-100 AB261724-1ML
ABCAAB261724-100EA 775 EUR
ABCAAB261724-100 ABCAAB261724-1ML
Anti-Cdk9 Rabbit Monoclonal Antibody [clone: EPR22956-60]
Anticorpi
Rabbit monoclonal [EPR22956-60] to Cdk9.
Attenzione: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Cdk9
Clonality: Monoclonal
Clone: EPR22956-60
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse;Human;Rat