Stampa…
Anti-Calcitonin receptor/CT-R Rabbit Monoclonal Antibody [clone: EPR27455-47]
Fornitore: Abcam
AB313335-100UL AB313335-10UL AB313335-20UL AB313335-1ML
ABCAAB313335-100ULEA 694 EUR
ABCAAB313335-100UL ABCAAB313335-10UL ABCAAB313335-20UL ABCAAB313335-1ML
Anti-Calcitonin receptor/CT-R Rabbit Monoclonal Antibody [clone: EPR27455-47]
Anticorpi
Rabbit monoclonal [EPR27455-47] to Calcitonin receptor/CT-R.
Type: Primary
Antigen: Calcitonin receptor/CT-R
Clonality: Monoclonal
Clone: EPR27455-47
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat