Stampa…

Anti-Thymidine Phosphorylase Rabbit Monoclonal Antibody [clone: EPR25629-129]

Fornitore: Abcam
AB284861-100 AB284861-20UL AB284861-1ML
ABCAAB284861-100EA 713 EUR
ABCAAB284861-100 ABCAAB284861-20UL ABCAAB284861-1ML
Anti-Thymidine Phosphorylase Rabbit Monoclonal Antibody [clone: EPR25629-129]
Anticorpi
Rabbit monoclonal [EPR25629-129] to Thymidine Phosphorylase.

Attenzione: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Thymidine Phosphorylase
Clonality: Monoclonal
Clone: EPR25629-129
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR