Stampa…

Anti-CD103 Rabbit Monoclonal Antibody [clone: EPR22590-27]

Fornitore: Abcam
AB224202-100 AB224202-1ML
ABCAAB224202-100EA 0 EUR
ABCAAB224202-100 ABCAAB224202-1ML
Anti-CD103 Rabbit Monoclonal Antibody [clone: EPR22590-27]
Anticorpi
Rabbit monoclonal [EPR22590-27] to CD103.

Attenzione: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: CD103
Clonality: Monoclonal
Clone: EPR22590-27
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR