Stampa…

Anti-LDL Receptor Rabbit Monoclonal Antibody [clone: EPR24874-56]

Fornitore: Abcam
AB271189-100 AB271189-20UL AB271189-1ML
ABCAAB271189-100EA 713 EUR
ABCAAB271189-100 ABCAAB271189-20UL ABCAAB271189-1ML
Anti-LDL Receptor Rabbit Monoclonal Antibody [clone: EPR24874-56]
Anticorpi
Rabbit monoclonal [EPR24874-56] to LDL Receptor.

Attenzione: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: LDL Receptor
Clonality: Monoclonal
Clone: EPR24874-56
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR