Stampa…
Anti-Proteasome 20S LMP7 Rabbit Monoclonal Antibody [clone: EPR14482(B)]
Fornitore: Abcam
AB180606-100 AB180606-1ML
ABCAAB180606-100EA 663 EUR
ABCAAB180606-100 ABCAAB180606-1ML
Anti-Proteasome 20S LMP7 Rabbit Monoclonal Antibody [clone: EPR14482(B)]
Anticorpi
Rabbit monoclonal [EPR14482(B)] to Proteasome 20S LMP7.
Attenzione: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Proteasome 20S LMP7
Clonality: Monoclonal
Clone: EPR14482(B)
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human