Stampa…

Anti-68kDa Neurofilament/NF-L Rabbit Monoclonal Antibody [clone: EPR22035-112]

Fornitore: Abcam
AB223343-100 AB223343-20UL AB223343-1ML
ABCAAB223343-100EA 0 EUR
ABCAAB223343-100 ABCAAB223343-20UL ABCAAB223343-1ML
Anti-68kDa Neurofilament/NF-L Rabbit Monoclonal Antibody [clone: EPR22035-112]
Anticorpi
Rabbit monoclonal [EPR22035-112] to 68kDa Neurofilament/NF-L.

Attenzione: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: 68kDa Neurofilament/NF-L
Clonality: Monoclonal
Clone: EPR22035-112
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR