Stampa…

Anti-Superoxide Dismutase 3/EC-SOD Rabbit Monoclonal Antibody [clone: EPR25619-120]

Fornitore: Abcam
AB289821-100 AB289821-1
ABCAAB289821-100EA 938 EUR
ABCAAB289821-100 ABCAAB289821-1
Anti-Superoxide Dismutase 3/EC-SOD Rabbit Monoclonal Antibody [clone: EPR25619-120]
Anticorpi
Rabbit monoclonal [EPR25619-120] to Superoxide Dismutase 3/EC-SOD - BSA and Azide free (Detector).

Attenzione: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Superoxide Dismutase 3/EC-SOD
Clonality: Monoclonal
Clone: EPR25619-120
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR