Stampa…

Anti-Kv1.1 potassium channel Mouse Monoclonal Antibody [clone: K20/78]

Fornitore: Abcam
AB254277-100 AB254277-1
ABCAAB254277-100EA 1020 EUR
ABCAAB254277-100 ABCAAB254277-1
Anti-Kv1.1 potassium channel Mouse Monoclonal Antibody [clone: K20/78]
Anticorpi
Mouse monoclonal [K20/78] to Kv1.1 potassium channel - BSA and Azide free.

Attenzione: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Kv1.1 potassium channel
Clonality: Monoclonal
Clone: K20/78
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Mouse;Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR