Stampa…

Anti-STAT6 Rabbit Monoclonal Antibody [clone: EPR18278-222]

Fornitore: Abcam
AB217998-100 AB217998-1ML
ABCAAB217998-100EA 707 EUR
ABCAAB217998-100 ABCAAB217998-1ML
Anti-STAT6 Rabbit Monoclonal Antibody [clone: EPR18278-222]
Anticorpi
Rabbit monoclonal [EPR18278-222] to STAT6.

Attenzione: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: STAT6
Clonality: Monoclonal
Clone: EPR18278-222
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR