Stampa…

Anti-PPAR gamma Rabbit Monoclonal Antibody [clone: EPR23297-111]

Fornitore: Abcam
AB272718-100 AB272718-1ML
ABCAAB272718-100EA 713 EUR
ABCAAB272718-100 ABCAAB272718-1ML
Anti-PPAR gamma Rabbit Monoclonal Antibody [clone: EPR23297-111]
Anticorpi
Rabbit monoclonal [EPR23297-111] to PPAR gamma.

Attenzione: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: PPAR gamma
Clonality: Monoclonal
Clone: EPR23297-111
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR