Stampa…

Anti-Histone H3 (citrulline R17) Rabbit Monoclonal Antibody [clone: EPR20358-120]

Fornitore: Abcam
AB232938-100 AB232938-1
ABCAAB232938-100EA 1020 EUR
ABCAAB232938-100 ABCAAB232938-1
Anti-Histone H3 (citrulline R17) Rabbit Monoclonal Antibody [clone: EPR20358-120]
Anticorpi
Rabbit monoclonal [EPR20358-120] to Histone H3 (citrulline R17) - BSA and Azide free.

Attenzione: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Histone H3
Clonality: Monoclonal
Clone: EPR20358-120
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR