Stampa…
Anti-Alanine Transaminase Rabbit Monoclonal Antibody [clone: EPR21068-102]
Fornitore: Abcam
AB244765-100 AB244765-1
ABCAAB244765-100EA 1040 EUR
ABCAAB244765-100 ABCAAB244765-1
Anti-Alanine Transaminase Rabbit Monoclonal Antibody [clone: EPR21068-102]
Anticorpi
Rabbit monoclonal [EPR21068-102] to Alanine Transaminase - BSA and Azide free (Capture).
Attenzione: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: Alanine Transaminase
Clonality: Monoclonal
Clone: EPR21068-102
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human