Stampa…

Anti-TCP1 delta Rabbit Monoclonal Antibody [clone: EPR8495(B)]

Fornitore: Abcam
AB248280-100 AB248280-1
ABCAAB248280-100EA 1020 EUR
ABCAAB248280-100 ABCAAB248280-1
Anti-TCP1 delta Rabbit Monoclonal Antibody [clone: EPR8495(B)]
Anticorpi
Rabbit monoclonal [EPR8495(B)] to TCP1 delta - BSA and Azide free.

Attenzione: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: TCP1 delta
Clonality: Monoclonal
Clone: EPR8495(B)
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR