Stampa…

Anti-PKC delta (phospho S645) Rabbit Monoclonal Antibody [clone: EP1486Y]

Fornitore: Abcam
AB284781-100 AB284781-1
ABCAAB284781-100EA 1020 EUR
ABCAAB284781-100 ABCAAB284781-1
Anti-PKC delta (phospho S645) Rabbit Monoclonal Antibody [clone: EP1486Y]
Anticorpi
Rabbit monoclonal [EP1486Y] to PKC delta (phospho S645) - BSA and Azide free.

Attenzione: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: PKC delta
Clonality: Monoclonal
Clone: EP1486Y
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR