Stampa…

Anti-CD62L Rabbit Monoclonal Antibody [clone: EPR17012-27]

Fornitore: Abcam
AB253240-100 AB253240-20UL AB253240-1ML
ABCAAB253240-100EA 738 EUR
ABCAAB253240-100 ABCAAB253240-20UL ABCAAB253240-1ML
Anti-CD62L Rabbit Monoclonal Antibody [clone: EPR17012-27]
Anticorpi
Rabbit monoclonal [EPR17012-27] to CD62L.

Attenzione: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: CD62L
Clonality: Monoclonal
Clone: EPR17012-27
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR