Stampa…

Anti-IL-5 Rabbit Monoclonal Antibody [clone: EPR16614-120]

Fornitore: Abcam
AB242707-100 AB242707-1
ABCAAB242707-100EA 938 EUR
ABCAAB242707-100 ABCAAB242707-1
Anti-IL-5 Rabbit Monoclonal Antibody [clone: EPR16614-120]
Anticorpi
Rabbit monoclonal [EPR16614-120] to IL-5 - BSA and Azide free (Detector).

Attenzione: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: IL-5
Clonality: Monoclonal
Clone: EPR16614-120
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR