Stampa…

Anti-BNP Rabbit Monoclonal Antibody [clone: EPR20260-222]

Fornitore: Abcam
AB309127-100UL AB309127-20UL AB309127-1ML
ABCAAB309127-100ULEA 694 EUR
ABCAAB309127-100UL ABCAAB309127-20UL ABCAAB309127-1ML
Anti-BNP Rabbit Monoclonal Antibody [clone: EPR20260-222]
Anticorpi
Rabbit monoclonal [EPR20260-222] to BNP.

Attenzione: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: BNP
Clonality: Monoclonal
Clone: EPR20260-222
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR