Stampa…
Anti-TMS1/ASC Rabbit Recombinant Multiclonal Antibody [clone: RM1049]
Fornitore: Abcam
AB309497-100UL AB309497-10UL AB309497-20UL AB309497-1ML
ABCAAB309497-100ULEA 582 EUR
ABCAAB309497-100UL ABCAAB309497-10UL ABCAAB309497-20UL ABCAAB309497-1ML
Anti-TMS1/ASC Rabbit Recombinant Multiclonal Antibody [clone: RM1049]
Anticorpi
Rabbit recombinant multiclonal [RM1049] to TMS1/ASC.
Attenzione: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: TMS1/ASC
Clonality: Recombinant Multiclonal
Clone: RM1049
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat