Stampa…
Anti-Glutamate Receptor 1 (AMPA subtype) Rabbit Monoclonal Antibody [clone: E308]
Fornitore: Abcam
AB247259-1 AB247259-100
ABCAAB247259-1EA 9160 EUR
ABCAAB247259-1 ABCAAB247259-100
Anti-Glutamate Receptor 1 (AMPA subtype) Rabbit Monoclonal Antibody [clone: E308]
Anticorpi
Rabbit monoclonal [E308] to Glutamate Receptor 1 (AMPA subtype) - BSA and Azide free.
Attenzione: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: Glutamate Receptor 1 (AMPA subtype)
Clonality: Monoclonal
Clone: E308
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human