Stampa…

Anti-CD55 Rabbit Monoclonal Antibody [clone: EPR6689] (HRP (Horseradish Peroxidase))

Fornitore: Abcam
AB309977-100UL AB309977-10UL
ABCAAB309977-100ULEA 819 EUR
ABCAAB309977-100UL ABCAAB309977-10UL
Anti-CD55 Rabbit Monoclonal Antibody [clone: EPR6689] (HRP (Horseradish Peroxidase))
Anticorpi
HRP Rabbit monoclonal [EPR6689] to CD55.

Attenzione: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: CD55
Clonality: Monoclonal
Clone: EPR6689
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR