Stampa…

Anti-CD16 Rat Monoclonal Antibody [clone: SP175]

Fornitore: Abcam
AB308607-100 AB308607-10 AB308607-20UL AB308607-1ML
ABCAAB308607-100EA 757 EUR
ABCAAB308607-100 ABCAAB308607-10 ABCAAB308607-20UL ABCAAB308607-1ML
Anti-CD16 Rat Monoclonal Antibody [clone: SP175]
Anticorpi
Rat monoclonal [SP175] to CD16 - Chimeric.

Attenzione: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: CD16
Clonality: Monoclonal
Clone: SP175
Conjugation:
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR