Stampa…
Anti-STAT1 (phospho S727) Rabbit Monoclonal Antibody [clone: Stat1S727-C6] (PE (Phycoerythrin))
Fornitore: Abcam
AB278719-10 AB278719-100
ABCAAB278719-10EA 269 EUR
ABCAAB278719-10 ABCAAB278719-100
Anti-STAT1 (phospho S727) Rabbit Monoclonal Antibody [clone: Stat1S727-C6] (PE (Phycoerythrin))
Anticorpi
PE Rabbit monoclonal [Stat1S727-C6] to STAT1 (phospho S727).
Attenzione: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: STAT1
Clonality: Monoclonal
Clone: Stat1S727-C6
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human