Stampa…

Anti-Polymeric immunoglobulin receptor/PIGR Rabbit Monoclonal Antibody [clone: EPR23314-78]

Fornitore: Abcam
AB272728-100 AB272728-1ML
ABCAAB272728-100EA 763 EUR
ABCAAB272728-100 ABCAAB272728-1ML
Anti-Polymeric immunoglobulin receptor/PIGR Rabbit Monoclonal Antibody [clone: EPR23314-78]
Anticorpi
Rabbit monoclonal [EPR23314-78] to Polymeric immunoglobulin receptor/PIGR.

Attenzione: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Polymeric immunoglobulin receptor/PIGR
Clonality: Monoclonal
Clone: EPR23314-78
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR