Stampa…
Anti-Collagen IV Rabbit Monoclonal Antibody [clone: EPR22911-127] (Alexa Fluor® 647)
Fornitore: Abcam
AB309503-100UL AB309503-10UL
ABCAAB309503-100ULEA 819 EUR
ABCAAB309503-100UL ABCAAB309503-10UL
Anti-Collagen IV Rabbit Monoclonal Antibody [clone: EPR22911-127] (Alexa Fluor® 647)
Anticorpi
Alexa Fluor® 647 Rabbit monoclonal [EPR22911-127] to Collagen IV.
Attenzione: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: Collagen IV
Clonality: Monoclonal
Clone: EPR22911-127
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat