Stampa…

Anti-GAS 6 Rabbit Monoclonal Antibody [clone: EPR23931-27]

Fornitore: Abcam
AB264098-100 AB264098-20UL AB264098-1ML
ABCAAB264098-100EA 713 EUR
ABCAAB264098-100 ABCAAB264098-20UL ABCAAB264098-1ML
Anti-GAS 6 Rabbit Monoclonal Antibody [clone: EPR23931-27]
Anticorpi
Rabbit monoclonal [EPR23931-27] to GAS 6.

Attenzione: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: GAS 6
Clonality: Monoclonal
Clone: EPR23931-27
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR