Stampa…

Anti-GPVI Rabbit Monoclonal Antibody [clone: EPR25690-60]

Fornitore: Abcam
AB305060-100 AB305060-20UL AB305060-1ML
ABCAAB305060-100EA 657 EUR
ABCAAB305060-100 ABCAAB305060-20UL ABCAAB305060-1ML
Anti-GPVI Rabbit Monoclonal Antibody [clone: EPR25690-60]
Anticorpi
Rabbit monoclonal [EPR25690-60] to GPVI.

Attenzione: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: GPVI
Clonality: Monoclonal
Clone: EPR25690-60
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR