Stampa…
Anti-Pro-Collagen III alpha 1 Rabbit Monoclonal Antibody [clone: EPR24255-13] (Alexa Fluor® 488)
Fornitore: Abcam
AB300097-10 AB300097-100
ABCAAB300097-10EA 238 EUR
ABCAAB300097-10 ABCAAB300097-100
Anti-Pro-Collagen III alpha 1 Rabbit Monoclonal Antibody [clone: EPR24255-13] (Alexa Fluor® 488)
Anticorpi
Alexa Fluor® 488 Rabbit monoclonal [EPR24255-13] to Pro-Collagen III alpha 1.
Attenzione: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: Pro-Collagen III alpha 1
Clonality: Monoclonal
Clone: EPR24255-13
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human