Stampa…

Anti-Galectin 10 Rabbit Monoclonal Antibody [clone: EPR11197]

Fornitore: Abcam
AB249338-100 AB249338-1
ABCAAB249338-100EA 1020 EUR
ABCAAB249338-100 ABCAAB249338-1
Anti-Galectin 10 Rabbit Monoclonal Antibody [clone: EPR11197]
Anticorpi
Rabbit monoclonal [EPR11197] to Galectin 10 - BSA and Azide free.

Attenzione: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Galectin 10
Clonality: Monoclonal
Clone: EPR11197
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR