Stampa…

Anti-VEGF Receptor 2 Rabbit Monoclonal Antibody [clone: EPR21884-236] (Alexa Fluor® 647)

Fornitore: Abcam
AB307529-100 AB307529-10
ABCAAB307529-100EA 819 EUR
ABCAAB307529-100 ABCAAB307529-10
Anti-VEGF Receptor 2 Rabbit Monoclonal Antibody [clone: EPR21884-236] (Alexa Fluor® 647)
Anticorpi
Alexa Fluor® 647 Rabbit monoclonal [EPR21884-236] to VEGF Receptor 2.

Attenzione: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: VEGF Receptor 2
Clonality: Monoclonal
Clone: EPR21884-236
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR