Stampa…

Anti-ERK1 Rabbit Monoclonal Antibody [clone: EP4967] (Alexa Fluor® 488)

Fornitore: Abcam
AB312910-100UL AB312910-10UL
ABCAAB312910-100ULEA 0 EUR
ABCAAB312910-100UL ABCAAB312910-10UL
Anti-ERK1 Rabbit Monoclonal Antibody [clone: EP4967] (Alexa Fluor® 488)
Anticorpi
Alexa Fluor® 488 Rabbit monoclonal [EP4967] to ERK1.

Attenzione: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: ERK1
Clonality: Monoclonal
Clone: EP4967
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR