Stampa…

Anti-Glutamate receptor 3/GluA3 Rabbit Monoclonal Antibody [clone: EP813Y]

Fornitore: Abcam
AB232430-100 AB232430-1
ABCAAB232430-100EA 1020 EUR
ABCAAB232430-100 ABCAAB232430-1
Anti-Glutamate receptor 3/GluA3 Rabbit Monoclonal Antibody [clone: EP813Y]
Anticorpi
Rabbit monoclonal [EP813Y] to Glutamate receptor 3/GluA3 - BSA and Azide free.

Attenzione: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: Glutamate receptor 3/GluA3
Clonality: Monoclonal
Clone: EP813Y
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR