Stampa…

Anti-Histone H2A+Histone H4 (symmetric di methyl R3) Rabbit Monoclonal Antibody [clone: EPR27303-14]

Fornitore: Abcam
AB309355-1MG AB309355-100UG
ABCAAB309355-1MGEA 9160 EUR
ABCAAB309355-1MG ABCAAB309355-100UG
Anti-Histone H2A+Histone H4 (symmetric di methyl R3) Rabbit Monoclonal Antibody [clone: EPR27303-14]
Anticorpi
Rabbit monoclonal [EPR27303-14] to Histone H2A (symmetric di methyl R3) + Histone H4 (symmetric di methyl R3) - BSA and Azide free.

Attenzione: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.

Type: Primary
Antigen: Histone H2A+Histone H4
Clonality: Monoclonal
Clone: EPR27303-14
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR